Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Biomek i3 Benchtop Liquid Handler

Biomek i3 Benchtop Liquid Handler

Biomek i3 Benchtop liquid handler, 1 - 1000 µl with air displacement technology
Catalog # 78641-438
New Product
Supplier:   Beckman Coulter
Supplier #:  D22660
Biomek i3 Benchtop Liquid Handler
Catalog # 78641-438
Supplier:  Beckman Coulter
New Product
Biomek i3 Benchtop Liquid Handler
Biomek i3 Benchtop Liquid Handler

Biomek i3 Benchtop liquid handler, 1 - 1000 µl with air displacement technology

Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The Biomek i3 Benchtop liquid handler is the newest and most compact member of the Biomek i-Series family, ideal for accelerating entry-level to medium-throughput workflows. It’s the perfect choice for those beginning their journey into lab automation or seeking a simple and affordable yet powerful alternative to more complex systems.

  • Simply empowering: No automation expertise required

  • Workflow enabling: Seamlessly adapts to your existing workflows
  • Space saving: Fits where you want it due to the small footprint
  • 15 ANSI/SLAS standard pipettable and five flex positions
  • Includes 12 standard and three short static automated labware positioners (ALPs)
  • Gripper supports labware up to 12.8 cm in height and 725 g in weight, and it's capable of stacking up to four plates
  • Ready for on-deck and off-deck integration (Left, Right, Back & Down)

Fixed-8 pipetting pod allows selective loading of 1 to 8 tips in any sequence. Uses standard Biomek i-Series 96-format disposable tips.

Key applications include Cell transfection, Nucleic acid extraction, ELISA, NGS library prep, Plasmid DNA prep, Antibody cocktailing, Mass spec sample prep, Cell passaging/media exchange, PCR/qPCR setup, AMPure XP cleanup, SPRIselect size selection, Valita Titer assay.

Windows® 11-compatible Biomek software with touchscreen support. Supports 21 CFR Part 11 with user accounts, permissions, and audit trails. BeckmanConnect enables secure remote diagnostics and troubleshooting. Guided labware setup. Method options selector step builder. Camera-enabled error log.

Specifications
  • Volume:
    1 - 1000 µl with air displacement technology
  • Description:
    Biomek i3 Benchtop liquid handler
  • Cat. No.:
    78641-438
  • W×D×H of system:
    73.2×78.5×82.6 cm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Biomek i3 service contract package
    Biomek i3 service contract package
    78641-440
    Log in for availability
    Each - $5,284.00