Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tiltable Arbor

Tiltable Arbor

Tiltable arbor, 25 mm (vertical post), 245 mm (vertical post)
Catalog # 78621-436
New Product
Supplier:   Aven Tools
Supplier #:  26800B-521
Tiltable Arbor
Catalog # 78621-436
Supplier:  Aven Tools
New Product
Tiltable Arbor

Tiltable arbor, 25 mm (vertical post), 245 mm (vertical post)

Price
$166.88
Log in for availability
Each
Product Details & Documents

Arbors are designed to extend the working range of boom stands and articulating arm stands. Allows tilting of microscope bodies to increase operator comfort. 25 mm arbor diameter, length 245 mm, and 15.875 mm (5/8") adapter.

  • Extends working range

  • Tiltable
  • Increases operator comfort

Ordering information: Stands sold separately.

Specifications
  • Description:
    Tiltable arbor
  • Length:
    245 mm (vertical post)
  • Diameter:
    25 mm (vertical post)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.