Orders LinkContactUsLinkComponent
Forceps - Serrated Tips
New Product
 :   Aven Tools
Forceps - Serrated Tips
New Product
Forceps - Serrated Tips
These serrated-tip long forceps, made of polished stainless steel, are ideal for retrieving parts from hard to access areas. Can also be used at home or kitchen for cooking, grilling, repairing, soldering etc. The length of this tool makes it ideal for the contact-less handling of objects.
  • Stainless steel finish provides excellent corrosion resistance
    Ideal for bending, separating and removing small items
    Long, straight tip is helpful when working in confined spaces
Applications
Accessing materials in confined or hard-to-reach areas
Handling components in electronics or laboratory settings
Cooking, grilling, and food preparation tasks
General-purpose use in crafts, hobbies, and repairs
Product Family Options
  • Description
    Material
    Tip length
    Tip style
    Weight
  • Forceps with straight serrated tips
    400 Stainless Steel
    200 mm (8")
    Straight Serrated
    45.36 g (0.1 lbs)
    Each
    Each - $8.76
    $8.76
    Each
       $8.76
     : 18434
    New Product
    New Product
  • Forceps with straight serrated tips
    400 Stainless Steel
    130 mm (5")
    Straight Serrated
    22.68 g (0.05 lbs)
    Each
    Each - $8.76
    $8.76
    Each
       $8.76
     : 18436
    New Product
    New Product
  • Splinter forceps with straight serrated tips
    400 Stainless Steel
    115 mm (4¹/₂")
    Straight Tip Serrated
    22.68 g (0.05 lbs)
    Each
    Each - $4.66
    $4.66
    Each
       $4.66
     : 18439
    New Product
    New Product
  • Forceps with straight serrated tips
    400 Stainless Steel
    300 mm (12")
    Straight Serrated
    90.72 g (0.2 lbs)
    Each
    Each - $11.56
    $11.56
    Each
       $11.56
     : 18432
    New Product
    New Product
  • Forceps with straight serrated tips
    400 Stainless Steel
    255 mm (10")
    Straight Serrated
    68.04 g (0.15 lbs)
    Each
    Each - $9.58
    $9.58
    Each
       $9.58
     : 18433