Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
pH buffers, colour coded, reference standards, Supelco®

pH buffers, colour coded, reference standards, Supelco®

New Product
Supplier:   MilliporeSigma
pH buffers, colour coded, reference standards, Supelco®
Supplier:  MilliporeSigma
New Product
Certificates
pH buffers, colour coded, reference standards, Supelco®
pH buffers, colour coded, reference standards, Supelco®

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Certifications: Standardized against N.I.S.T. standard reference materials

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 78607-858
  • 78607-220
  • M-Clarity quality level:
  • MQ100
  • MQ100

Specifications

Product Family Options

Product Information

  • Reference temperature
    Size
    pH
    Accuracy
    Color
    Availability
    Price
  • 25 °C
    3.8 L (1 gal.)
    7.00
    ±0.01
    Yellow
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $280.00
    $280.00
    Each(1/Itm/Bx)
       $280.00

    Supplier Number: B4770-1GA
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78607-858
    M-Clarity quality level
    MQ100
    New Product
  • 25 °C
    6×500 ml
    10.00
    ±0.01
    Blue
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $264.00
    $264.00
    Each(1/Itm/Bx)
       $264.00

    Supplier Number: B4895-6X500ML
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78607-220
    M-Clarity quality level
    MQ100
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.