Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Buffer, Reference Standard pH 1.68 ±0.01 at 25 °C

Buffer, Reference Standard pH 1.68 ±0.01 at 25 °C

New Product
Supplier:   BDH
Buffer, Reference Standard pH 1.68 ±0.01 at 25 °C
Supplier:  BDH
New Product
Certificates
Buffer, Reference Standard pH 1.68 ±0.01 at 25 °C

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • pH at 25 °C (Method: SQCP027, SQCP033): 1.67 to 1.69

CAS:7732-18-5,6100-20-5

Product Family Options

Product Information

  • pH accuracy
    Packed
    Size
    pH
    Color
    Storage temperature
    Availability
    Price
  • ±0.01 at 25 °C
    Pouches
    20×20 ml
    1.68
    Colourless
    Ambient
    Log in for availability
    Log in for availability
    Pack of 20 (20ml)
    Pack of 20 (20ml) - $78.25
    $78.25
    Pack of 20 (20ml)
       $78.25

    Specifications:
    New Product
    New Product
  • ±0.01 at 25 °C
    Poly bottle
    1 L
    1.68
    Colourless
    Ambient
    Log in for availability
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $48.19
    $48.19
    Log in for availability
    Case of 12 (1/ Itm/Bx)
    Case of 12 (1/ Itm/Bx) - $922.50
    $922.50
    Each(1/Itm/Bx)
       $48.19

    Case of 12 (1/ Itm/Bx)
       $922.50

    Specifications:
    New Product
    New Product
  • ±0.01 at 25 °C
    Cubitainer®
    20 L
    1.68
    Colourless
    Ambient
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $160.69
    $160.69
    Each(1/Itm/Bx)
       $160.69

    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.