Orders LinkContactUsLinkComponent
Pipette Controller ali-Q® Edge
ali-Q® Edge
  78535-088
New Product
 :  2300-3005
Pipette Controller ali-Q® Edge
  78535-088
New Product
Pipette Controller ali-Q® Edge
Pipette Controller ali-Q® Edge
ali-Q® Edge
$707.94
Case of 1
The ali-Q® Edge aliquoting pipet controller delivers accurate repeat dispenses, eliminating the need to watch the graduation lines on serological pipets.
  • Enter precise aliquot dispense volumes using digital display
  • Universally accepts any serological pipet to consistently deliver aliquots from 0.5 to 5.0 ml
  • Compact size and intuitive design allows easy toggling between traditional dispense and aliquot modes
  • LEDs built into the translucent air tank clearly indicate dispense mode and battery/charging status from any viewing angle
  • The ali-Q Edge® is self-standing, or may be used with included and removable kickstand
  • Lithium ion battery recharges via USB-C port
  • Description:
    ali-Q® Edge
Accessories
  •  
    Description
  • 78535-140
    Nozzle cover for ali-Q® Edge
    Case of 1 - $51.20
  •  
    Description
  • 78535-092
    Battery cover for ali-Q® Edge
    Case of 1 - $43.09
  •  
    Description
  • 78535-102
    Kickstand for ali-Q® Edge
    Case of 1 - $33.86