Orders LinkContactUsLinkComponent
Sealing Ring
Sealing Ring
Sealing ring, 40.1H×184.9Ø mm
  78474-021
New Product
 :   SAVILLEX
Supplier #:  550-740-400823
Sealing Ring
  78474-021
 :  SAVILLEX
New Product
Sealing Ring
Sealing ring, 40.1H×184.9Ø mm
$277.86
Each
Minimum order quantity is 1
Fits cleaning container and silicone grip mat (78473-766) is recommended for tightening and loosening the sealing ring.
  • Upper service temperature of ECTFE is 150 °C (300 °F)
  • Description:
    Sealing ring
  • Dimensions:
    40.1H×184.9Ø mm