Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Storage Containers with Optional Lid

Storage Containers with Optional Lid

Supplier:   Mopec
Storage Containers with Optional Lid
Supplier:  Mopec
Certificates
Storage Containers with Optional Lid
Storage Containers with Optional Lid

2 Options Available Below

Product Details & Documents

Ideal for dissection instruments and utensils storage. Flush handle allows stacking.

  • Type 304 stainless steel

  • Dimensions, Diameter×H: 160×170 mm (6.5×7") 
  • Capacity: 3 L (3.1875 quarts)

The Mopec Stainless Steel Storage Container with optional lid offers a durable and hygienic solution for storing, transporting, and organizing instruments or specimens in laboratory and pathology environments. Crafted from high-quality stainless steel, this container provides exceptional resistance to corrosion, staining, and chemical exposure, ensuring long-lasting performance in demanding conditions.

Product Family Options

Product Information

  • Capacity
    Dimensions
    Availability
    Price
  • 76549-558
    Storage container, stainless steel
    Storage container, stainless steel
    3.1875 quarts
    6.5Ø×7"H
    Log in for availability
    Log in for availability
    Each
    Each - $132.56
    $132.56
    Each
       $132.56

    Supplier Number: BA124
  • 76549-560
    Storage container lid, stainless steel
    Storage container lid, stainless steel
    Ø6.5"
    Log in for availability
    Log in for availability
    Each
    Each - $59.91
    $59.91
    Each
       $59.91

    Supplier Number: BA125