Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hemicholinium-3 ≥95%, Moligand™

Hemicholinium-3 ≥95%, Moligand™

Irritant
Toxic
New Product
Supplier:   Aladdin Scientific
Hemicholinium-3 ≥95%, Moligand™
Supplier:  Aladdin Scientific
New Product
Certificates
Hemicholinium-3 ≥95%, Moligand™

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A potent and selective choline uptake blocker.

Synonyms:  Hemicholine, 2,2′-(4,4′-Biphenylene)bis(2-hydroxy-4,4-dimethylmorpholinium bromide)
  • CAS Number:312-45-8
  • Formula:C₂₄H₃₄Br₂N₂O₄
  • Molecular Weight:574.35 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:206-227-3
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    10 mg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $78.90
    $78.90
    Each(1/Itm/Bx)
       $78.90

    Supplier Number: H346775-10MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • Glass bottle
    25 mg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $178.00
    $178.00
    Each(1/Itm/Bx)
       $178.00

    Supplier Number: H346775-25MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • Glass bottle
    5 mg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $51.90
    $51.90
    Each(1/Itm/Bx)
       $51.90

    Supplier Number: H346775-5MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.