Orders LinkContactUsLinkComponent
Nylon 6, rod, 15 mm
New Product
 :   Goodfellow
Nylon 6, rod, 15 mm
New Product
Nylon 6, rod, 15 mm
We offer a wide range of Nylon 6 Rods with 87 variations to choose from. These rods are made from an engineering plastic with good abrasion resistance, low friction, and the ability to flex without fracturing. Potential applications include bearings, wear pads, rollers and bushings. With their toughness, low weight and self-lubricating properties, Nylon 6 Rods provide versatile solutions for the manufacturing, automotive and industrial sectors.
  •  :PA 6 - (C₆H₁₁N)ₙ
Product Family Options
  • Size
  • Length 1000 mm
    Each
    Each - $217.00
    $217.00
    Each
       $217.00
     : 1000026946
    New Product
    New Product
  • Length 100 mm
    Each
    Each - $202.00
    $202.00
    Each
       $202.00
     : 1000026948
    New Product
    New Product
  • Length 10 mm
    Each
    Each - $199.00
    $199.00
    Each
       $199.00
     : 1000026952
    New Product
    New Product
  • Length 2000 mm
    Each
    Each - $230.00
    $230.00
    Each
       $230.00
     : 1000026955
    New Product
    New Product
  • Length 200 mm
    Each
    Each - $204.00
    $204.00
    Each
       $204.00
     : 1000026959
    New Product
    New Product
  • Length 20 mm
    Each
    Each - $199.00
    $199.00
    Each
       $199.00
     : 1000026962
    New Product
    New Product
  • Length 3000 mm
    Each
    Each - $241.00
    $241.00
    Each
       $241.00
     : 1000026965
    New Product
    New Product
  • Length 300 mm
    Each
    Each - $206.00
    $206.00
    Each
       $206.00
     : 1000026969
    New Product
    New Product
  • Length 30 mm
    Each
    Each - $200.00
    $200.00
    Each
       $200.00
     : 1000026973
    New Product
    New Product
  • Length 500 mm
    Each
    Each - $209.00
    $209.00
    Each
       $209.00
     : 1000026977
    New Product
    New Product
  • Length 50 mm
    Each
    Each - $200.00
    $200.00
    Each
       $200.00
     : 1000026980