Orders LinkContactUsLinkComponent
STEMdiff™ Forebrain Neuron Differentiation Kit
STEMdiff™ Forebrain Neuron Differentiation Kit
STEMdiff™ Forebrain neuron differentiation kit, 1 kit
Catalog # MSPP-8600
Marketsource
Supplier #:  8600
STEMdiff™ Forebrain Neuron Differentiation Kit
Catalog # MSPP-8600
Marketsource
CAS Number:  
STEMdiff™ Forebrain Neuron Differentiation Kit
STEMdiff™ Forebrain Neuron Differentiation Kit
STEMdiff™ Forebrain neuron differentiation kit, 1 kit
CAS Number:  
Price
$573.85
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Differentiation kit for the generation of neuronal precursors from human ES and iPS cell-derived neural progenitor cells.
The STEMdiff™ Forebrain Neuron Differentiation Kit is used in conjunction with the STEMdiff™ Forebrain Neuron Maturation Kit to generate a mixed population of forebrain-type (FOXG1-positive) neurons from neural progenitor cells derived from human pluripotent stem cells. This kit is optimised to work with STEMdiff™ SMADi Neural Induction Kit, which supplies the appropriate neural progenitor cells. Neurons derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Description:
    STEMdiff™ Forebrain neuron differentiation kit
  • Size:
    1 kit