Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Litesizer, DIF 500

Litesizer, DIF 500

Litesizer DIF 500
Catalog # 78172-174
New Product
Supplier:   Anton Paar
Supplier #:  261784
Litesizer, DIF 500
Catalog # 78172-174
Supplier:  Anton Paar
New Product
Litesizer, DIF 500
Litesizer, DIF 500

Litesizer DIF 500

Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Litesizer DIF 500 performs particle size analysis from 10 nm to 3.5 mm via laser diffraction, utilizing dual 10 mW and 25 mW lasers and angular detection from 0.01°–170°. Kalliope software enables three-click measurements. Quick-Click dispersion switching, integrated safety protocols, and a vibration-isolated optical bench ensure precision in demanding environments.

  • Measure samples from 0.01 µm to 3500 µm
  • Switch between liquid and dry dispersion in just one move
  • Kalliope software: No training required, dedicated QC mode
  • Reliable results even in harsh environments

Litesizer DIF 500 Specifications Measurement principle: Laser Diffraction (Mie and Fraunhofer scattering) Measurement range: 0.01 to 3,500 μm Size classes: 144 (user adjustable) Accuracy: better than ± 0.5 % variation+ Repeatability: better than ± 0.5 % variation+ Reproducibility: better than ± 1 % variation+ Possible measurement duration: <10 sec Data acquisition rate: 16 kHz LIGHT SOURCE 1 Type: Fiber coupled laser diode Optical arrangement: Reverse Fourier Wave length: 830 nm, infrared Power: 10 mW Laser class: Class 1 (IEC60825-1) LIGHT SOURCE 2 Type L: Laser diode Optical arrangement: Tilted relative to IR laser Wave length: 450 nm, blue Power: 25 mW Laser class: Class 1 (IEC60825-1) DETECTORS Type: Log-spaced photo diode array and single diodes for side and back-scattering Angular range: 0.01° to 170° Focal length: 300 mm Alignment: automatic INSTRUMENT SIZE Dimensions: 400 mm x 790 mm x 290 mm (H x W x D) Weight: 42.3 kg (93.2 lb) Power supply: 100 V to 240 V ±10 %, 50/60 Hz OPERATING CONDITIONS Temperature: 10 °C to 30 ºC Humidity: 35 % to 80 % non-condensing IP rating: IP 41

Certifications: Compliant with standards: ISO 13320:2020, USP 429, ASTM B822 – 20, ASTM D4464 - 15(2020), ASTM E2316 - 14(2019).

Specifications
  • Description:
    Litesizer DIF 500
Accessories

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Particle size standard PS 415
    Particle size standard PS 415
    78172-196
    Particle size standard for validating with Dry jet dispersion unit
    Log in for availability
    Each - $860.30

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software 21 CFR PART 11
    Kalliope software 21 CFR PART 11
    78172-102
    Software module for Kalliope to comply with the regulation 21 CFR Part 11 introduced by US Food and Drug Administration (FDA);Key features: electronic signature, audit trail, retrieval
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software professional
    Kalliope software professional
    78172-076
    One-page workflow displays all measurement details in one sight, including input parameters, results, and expert advice;Meets US FDA Regulation 21 CFR Part 11 standards for electronic records, audit trails, and signatures when used in conjunction with the KALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11;Includes license for 3 activations (installations)
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Sample funnel, large
    Sample funnel, large
    78172-158
    Large funnel (max. 600 ml) serving as a reservoir for the sample right before the measurement; Compatible with Dry Jet dispersion unit and Free Fall dispersion unit.
    Log in for availability
    Each - $770.72

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software unlimitted
    Kalliope software unlimitted
    78172-078
    Measurement series, databases for solvents, materials and SOPs are all easily accessible;Results from separate experiments can be compared and analyzed, and presented in customizable reports and/or exported to an Excel file for further processing and analysis;In combination with KALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11 the software complies with the US FDA’s Regulation 21 CFR Part 11 concerning electronic records, audit trails and signatures; license for unlimited installations
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Dry jet dispersion unit
    Dry jet dispersion unit
    78172-178
    Suitable for particles of up to 3500 µm (default);Optional: up to 5000 µm;Pressure: 0.05 - 4.6 bar; Sample funnel volume: 150 ml (default). Optional: 600 ml;Superior protection against sample spread;Automated feeding rate; Requires vacuum cleaner for operation;Minimum requirements to run: Supply of compressed gas 5 - 10 bar;Compatible vacuum cleaner (No. 184481, No. 188973, No. 188974 or No. 187511)
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Free fall dispersion unit
    Free fall dispersion unit
    78172-180
    Suitable for free-flowing and non-agglomerating particles up to 16000 µm (sample dependent);Measurement cell: 4 mm gap (default), Optional: 8 mm gap;Sample funnel volume: 600 ml (default), Optional: 150 ml;Superior protection against sample spread;Automated feeding rate; The sample is retrieved after the measurement
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Liquid flow dispersion unit
    Liquid flow dispersion unit
    78172-176
    Flexible working volume: 150 - 600 ml;Automated filling, rinsing, and draining;Minimum requirements to run: outflow of the liquid (wastewater water connection or a tank); Recommended requirements to run: water supply 0.1 - 8 bar
    Log in for availability
    Each

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.