Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Dynamic Image Analyzer, DIA 700

DIA 700 Dynamic image analyzer
Catalog # 78172-172
New Product
Supplier:   Anton Paar
 :  261782
Dynamic Image Analyzer, DIA 700
Catalog # 78172-172
Supplier:  Anton Paar
New Product
Dynamic Image Analyzer, DIA 700
Dynamic Image Analyzer, DIA 700

DIA 700 Dynamic image analyzer

Price
Inquire For Price
Log in for availability
Each
 
Product Details & Documents

The Litesizer DIA Series dynamic image analyzer characterizes particle size and shape through direct imaging. Easily switch between liquid, air, or free-fall dispersion modules in one step. Automated feed rate adjustment and liquid rinsing streamline operation. Minimal training is needed for fast, reliable measurements and efficient sample handling.

  • Triple magnification for maximum measurement precision
  • Analyze 0.5 µm to 16,000 µm samples with single particle resolution
  • Streamline your workflow with user-friendly Kalliope software
  • Automate tasks, freeing up valuable time for critical work
  • Compatible with Liquid Flow, Dry Jet, Free Fall units

Specifications Litesizer DIA 700 Measurement Principle: Dynamic Image Analysis Measurement Ranges (Liquid Flow): 0.5 to 2,500 μm Measurement Ranges (Dry Jet): 0.5 μm to 5,000 μm Measurement Ranges (Free Fall): 0.5 μm to 16,000 μm (restrictions apply for particles >8,000 μm) Camera: 5 Mpix (2,448 pixels x 2,048 pixels) / 0.5 μm per pixel Data Collection Rate 220 fps/camera at 5 Mpix Magnification: 0.3x, 1x and 6x Optical Features: Automatic switch between objectives Automatic merge of size ranges All magnifications are included in the standard configuration - Data Transfer: 1x 10 Gigabit Ethernet, 1x USB-A 3.0 Measurement Output Number-, surface-, and volume-weighted results Size descriptors (ISO 9276-compliant): Minimum and maximum Feret diameters, projected area equivalent diameter, length, geodesic length and thickness (e.g., for fibers), minimum and maximum axes of the Legendre ellipse Shape descriptors (ISO 9276-compliant): aspect ratio, ellipse ratio, irregularity, elongation or eccentricity, circularity, form factor, compactness, extent or bulkiness, solidity, convexity Image parameters: sharpness and contrast Instrument Data Dimensions: 400 mm x 790 mm x 290 mm (H x W x D) * Weight: 41 kg (90 lb) Power supply: 100 V to 240 V ±10 %, 50/60 Hz Compressed air supply: 5 bar - 10 bar (72 psi to 145 psi) Water supply: max. 8 bar (116 psi) Compatible Dispersion Units Liquid Flow: Dispersion using a liquid carrier, mixing, and ultrasonication Dry Jet: Dispersion via compressed air Free Fall: Dispersion via gravitational fall Exchange of the Dispersion Unit Less than 10 seconds via Quick Click Automation Automatic adjustment of the image acquisition rate Accessories Calibration tool Pre-configured PC

Specifications
  • Description:
    DIA 700 Dynamic image analyzer
Accessories

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Particle size standard PS 413
    Particle size standard PS 413
    78172-190
    Particle size standard for validating Zoom and Standard;Objectives with Liquid Flow dispersion unit
    Log in for availability
    Each - $465.23

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software professional
    Kalliope software professional
    78172-076
    One-page workflow displays all measurement details in one sight, including input parameters, results, and expert advice;Meets US FDA Regulation 21 CFR Part 11 standards for electronic records, audit trails, and signatures when used in conjunction with the KALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11;Includes license for 3 activations (installations)
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software 21 CFR PART 11
    Kalliope software 21 CFR PART 11
    78172-102
    Software module for Kalliope to comply with the regulation 21 CFR Part 11 introduced by US Food and Drug Administration (FDA);Key features: electronic signature, audit trail, retrieval
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Vacuum cleaner, 110 V
    Vacuum cleaner, 110 V
    78172-094
    Necessary accessory for Litesizer DIA with Dry Jet dispersion unit: collects measured powder samples at the end of the air jetstream;110 V A/C;Non-ATEX: not suitable for measuring flammable powders; Dust class H – highly hazardous dusts;Delivered with 15 A/240 V adapter cable for connection to the instrument;Disposable HEPA filter bags available (No. 199002); ATTENTION: The adapter cable should only be used to connect the vacuum cleaner to the instrument. Never connect the vacuum cleaner to the mains using this adapter!
    Log in for availability
    Each - $1,511.20

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Diffuser set 5 mm dry
    Diffuser set 5 mm dry
    78172-182
    Venturi tube of 5 mm diameter and 5 mm nozzle forming a stream of particles in Dry Jet dispersion unit;Use this diffuser if at least a fraction of the sample has a particle size between 3.5 mm and 5 mm.
    Log in for availability
    Each - $1,400.01

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Particle size standard PS 414
    Particle size standard PS 414
    78172-194
    Particle size standard for validating Zoom and Standard objectives with Dry Jet dispersion unit
    Log in for availability
    Each - $670.33

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Water filter kit
    Water filter kit
    78172-100
    Contains following separate materials: 1# pressure reducing gauge 1.5-6 bar, 1# fine filter holder P3, 1# fine filter cartridge P3, 1# coarse filter housing LA 1T, 1# coarse filter cartridge LA 1T, 1# ball valve, Various connectors, Inlet connection to water filter kit: ³/₄" external thread, Size: 62×16×40 cm (l/w/h); ATTENTION: Installation needs to be performed by a licensed plumber; NOTE: Replacement filter cartridges are available (Mat. 198484, 198486)
    Log in for availability
    Each - $2,439.50

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Protective glass for camera unit
    Protective glass for camera unit
    78172-154
    Protective glass 50.8×3 mm for the camera window in the main unit
    Log in for availability
    Each - $33.46

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Particle size standard ps 226
    Particle size standard ps 226
    78172-104
    Particle size standard for validating Zoom objective with Free Fall dispersion unit
    Log in for availability
    Each - $718.89

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Protective glass for illumination unit
    Protective glass for illumination unit
    78172-152
    Protective glass 76.2×3 mm for the illumination unit in the main unit
    Log in for availability
    Each - $33.46

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Sample funnel small
    Sample funnel small
    78172-156
    Small funnel (max. 150 ml) serving as a reservoir for the sample right before the measurement. Compatible with Dry Jet dispersion unit and Free Fall dispersion unit.
    Log in for availability
    Each - $591.53

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Sample and cleaning kit
    Sample and cleaning kit
    78172-188
    Includes Double spatula, Bottle brush, Q-Tips extra long, Cleaning brush, Microfiber cloth, Lens paper, Spoon 26×15 mm
    Log in for availability
    Each - $102.55

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Textile cover for dispersion unit
    Textile cover for dispersion unit
    78172-186
    Textile cover for protecting the dispersion unit while not in use
    Log in for availability
    Each - $85.27

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Calibration tool dia
    Calibration tool dia
    78172-166
    Required if the user wants to autonomously calibrate the instrument. For service technicians, the item is included in Mat. No. 264541 INSTALLATION AND MAINTENANCE KIT FOR LITESIZER DIA.
    Log in for availability
    Each - $3,880.54

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Air dryer IDG30 with air filter unit
    Air dryer IDG30 with air filter unit
    78172-164
    To separate oil, particle and condensate with fine filter 0.3 µm and micro filter 0.01 µm;dew point at outlet: –40 °C volume rate at outlet: 75 l/min;working pressure: 0.5 - 10 bar;NOTE: Replace CLEARPOINT fine filter (Mat. No. 11242) and subtlest filter (Mat. No. 11243) every 6 months
    Log in for availability
    Each - $1,839.34

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Particle size standard ps 234
    Particle size standard ps 234
    78172-192
    Particle size standard for validating Standard objectives with Free Fall dispersion unit
    Log in for availability
    Each - $626.07

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Liquid flow dispersion unit
    Liquid flow dispersion unit
    78172-176
    Flexible working volume: 150 - 600 ml;Automated filling, rinsing, and draining;Minimum requirements to run: outflow of the liquid (wastewater water connection or a tank); Recommended requirements to run: water supply 0.1 - 8 bar
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • ATEX approved vacuum cleaner, 110 V
    ATEX approved vacuum cleaner, 110 V
    78172-096
    Collects powder samples after the measurement at the end of the air jet stream;Suitable for collecting potentially explosive and flammable powder samples;Suitable for collecting powder samples hazardous to health;Dust class H – highly hazardous dusts;Motor: 110 V, 1150 W max, 1000 W mean;Performance: 59 L/sec, 2300 mm/WG;Dimensions: 830×440×440 mm, 21.2 kg
    Log in for availability
    Each - $6,098.77

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software unlimitted
    Kalliope software unlimitted
    78172-078
    Measurement series, databases for solvents, materials and SOPs are all easily accessible;Results from separate experiments can be compared and analyzed, and presented in customizable reports and/or exported to an Excel file for further processing and analysis;In combination with KALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11 the software complies with the US FDA’s Regulation 21 CFR Part 11 concerning electronic records, audit trails and signatures; license for unlimited installations
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Measurement cell 8 mm free fall
    Measurement cell 8 mm free fall
    78172-184
    Measurement cell for Free Fall dispersion unit with 8 mm gap;Compatible with Free Fall dispersion unit
    Log in for availability
    Each - $1,604.02

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Free fall dispersion unit
    Free fall dispersion unit
    78172-180
    Suitable for free-flowing and non-agglomerating particles up to 16000 µm (sample dependent);Measurement cell: 4 mm gap (default), Optional: 8 mm gap;Sample funnel volume: 600 ml (default), Optional: 150 ml;Superior protection against sample spread;Automated feeding rate; The sample is retrieved after the measurement
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Dry jet dispersion unit
    Dry jet dispersion unit
    78172-178
    Suitable for particles of up to 3500 µm (default);Optional: up to 5000 µm;Pressure: 0.05 - 4.6 bar; Sample funnel volume: 150 ml (default). Optional: 600 ml;Superior protection against sample spread;Automated feeding rate; Requires vacuum cleaner for operation;Minimum requirements to run: Supply of compressed gas 5 - 10 bar;Compatible vacuum cleaner (No. 184481, No. 188973, No. 188974 or No. 187511)
    Log in for availability
    Each