Orders LinkContactUsLinkComponent
Base Unit, Lovis 2001
Lovis 2001 Base Unit
  78454-860
New Product
 :   Anton Paar
 :  314855
Base Unit, Lovis 2001
  78454-860
New Product
Base Unit, Lovis 2001
Base Unit, Lovis 2001
Lovis 2001 Base Unit
Each
Lovis 2001 Base Unit to operate Lovis 2001 Microviscometer Module as independent instrument without combined density meter: Lovis 2001 Microviscometer = Lovis 2001 Microviscometer Module (model 314856) + Lovis 2001 Base Unit (model 314855).
  • Falling ball viscometer with adjustable angle fitting sample’s viscosity
  • Delivers intrinsic viscosity, K-value, average molar mass and more
  • Referenced in US Pharmacopoeia chapter 913 and Ph. Eur. 2.2.49
  • Automated filling and cleaning via sample changer
  • Combine to measure refractive index and pH in one go
MEASURING RANGE Dynamic viscosity: 0.2 to 10,000 mPa·s Shear rate: 0.5 to 1 to 1,000 s⁻¹ Temperature; 5 °C to 100 °C (41 °F to 212 °F) -40 °C to +100 °C (-40 °F to +212 °F) (with counter cooling)1)
PRECISION Viscosity: Repeatability s.d.: 0.1 % 2) Viscosity: Accuracy: 0.5 % 3) Measuring time: Resolution: 0.001 s Measuring time: Accuracy: 0.05 % Temperature: Repeatability s.d.: 0.005 °C Temperature: Accuracy: 0.02 °C Inclination: Repeatability s.d.: 0.02° Inclination: Accuracy: 0.1°
  • Description:
    Lovis 2001 Base Unit
Accessories
  •  
    Description
  • 78454-796
    Viscosity standard H20, 1000 ml
    Each - $133.26
  •  
    Description
  • 78454-894
    Viscosity standard H20, 100 ml certified
    Each - $132.19
  •  
    Description
  • 78454-832
    Accessory kit complete - lovis
    Each - $5,759.45
  •  
    Description
  • 78454-996
    Option lovis 2000 m/me low temperature
    Each - $5,273.33