Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dynamic Light Scattering Instrument, Litesizer DLS 101

Dynamic Light Scattering Instrument, Litesizer DLS 101

Dynamic light scattering instrument, Litesizer DLS 101
Catalog # 78172-202
New Product
Supplier:   Anton Paar
Supplier #:  317783
Dynamic Light Scattering Instrument, Litesizer DLS 101
Catalog # 78172-202
Supplier:  Anton Paar
New Product
Dynamic Light Scattering Instrument, Litesizer DLS 101
Dynamic Light Scattering Instrument, Litesizer DLS 101

Dynamic light scattering instrument, Litesizer DLS 101

Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Litesizer DLS enables high-resolution particle sizing via MAPS multi-angle dynamic light scattering and automatic angle optimization. Continuous transmittance monitoring detects sedimentation and agglomeration in real time. cmPALS and Omega Cuvette enhance zeta potential precision by mitigating aging and field distortion. Fluorescence/polarization filters support concentration analysis across MAPS modes.

  • Particle sizing from 0.3 nm to 10 µm
  • Cost-effective entry to dynamic light scattering
  • DLS back-scatter detection ensures broad sample coverage
  • Transmittance measurement to live-monitor sample behavior

Litesizer DLS 101 specifications:
Measurement principle: Dynamic Light Scattering (DLS)
Measuring range: 0.3 nm to 10 μm* (particle diameter)
Measurement angles: 175°
Min. concentration: 0.1 mg/ml (lysozyme)
Max. concentration: 50% w/v (sample-dependent)
Min. sample volume: 12 µl
Accuracy: Better than ±2% on NIST traceable standards
Repeatability: Better than ±2% on NIST traceable standards

Transmittance specifications:
Measuring range: 0 to 100%
Min. sample volume: 15 µl
Accuracy: Better than ±1%

General specifications:
Light source: Semiconductor laser diode/40 mW, 658 nm
Laser warm-up time: 6 min
Temperature control range: 0 to 120 °C
Ambient operating temperature: 10 to 35 °C
Humidity: Up to 80% non-condensing
Dimensions: 450W×505D×135H mm
Weight: Approx. 18 kg (40 lbs.)

Specifications
  • Description:
    Dynamic light scattering instrument, Litesizer DLS 101
Accessories

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Litesizer DLS filter exchange tool, spare part
    Litesizer DLS filter exchange tool, spare part
    78172-216
    For easy exchange of fluorescence- and polarization filters
    Log in for availability
    Each - $157.59

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Low volume configuration
    Low volume configuration
    78172-220
    Log in for availability
    Each - $3,781.24

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Flow module FM 10
    Flow module FM 10
    78172-206
    Every Litesizer DLS needs to be paired with a new BM11/FM11 once by a service technician; If the module is ordered together with the Litesizer DLS, this is done at the factory
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software 21 CFR PART 11
    Kalliope software 21 CFR PART 11
    78172-102
    Software module for Kalliope to comply with the regulation 21 CFR Part 11 introduced by US Food and Drug Administration (FDA);Key features: electronic signature, audit trail, retrieval
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Quartz cuvette; Outer dimensions: 12.5×12.5×45 mm; light path 10 mm; Not compatible with Univette (Cat. No: 78172-090)
    Quartz cuvette; Outer dimensions: 12.5×12.5×45 mm; light path 10 mm; Not compatible with Univette (Cat. No: 78172-090)
    78172-148
    Log in for availability
    Each - $234.23

Product Information

  • Catalog #
    Details
    Availability
    Price
  • TI stand 804
    TI stand 804
    78172-118
    Titration stand and controller for the rod stirrer 802; Together with the rod stirrer 802, it provides an addition to the magnetic stirrer forthe dosing system; The titration stand can be directly connected via the MSB connection cable instead of the magnetic stirrer
    Log in for availability
    Each - $1,321.22

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Syringe filter - ANOTOP 25
    Syringe filter - ANOTOP 25
    78172-068
    Non sterile, 0.02 µm membrane filter, Ø 20 mm, (1 piece)
    Log in for availability
    Each - $39.93

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Disposable cuvette lids
    Disposable cuvette lids
    78172-146
    One pack of 100 pieces
    Log in for availability
    Pack of 100 - $56.13

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Rod stirrer 802
    Rod stirrer 802
    78172-120
    Together with Ti stand 804 provides an addition to the magnetic stirrer of the dosing system
    Log in for availability
    Each - $1,110.74

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Batch module BM 10
    Batch module BM 10
    78172-204
    Every Litesizer DLS needs to be paired with a new BM11/FM11 once by a service technician; If the module is ordered together with the Litesizer DLS, this is done at the factory
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Low-volume disposable-cuvette set
    Low-volume disposable-cuvette set
    78172-082
    Pack of 80 disposable 50 L cuvettes plus one cuvette lifter; Outer dimensions: 12.5×12.5×36 mm; 2 light paths available 10 mm and 2 mm; for both light paths; side scattering only
    Log in for availability
    Each - $210.49

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Dosing system accessory DLS 701/501
    Dosing system accessory DLS 701/501
    78172-088
    Performs automated adjustment of the pH in the flow cuvette and enables pH ramp measurements
    Log in for availability
    Each - $21,545.37

Product Information

  • Catalog #
    Details
    Availability
    Price
  • NIST Traceable standard, 0.002% (15 ml), 220 nm
    NIST Traceable standard, 0.002% (15 ml), 220 nm
    78172-116
    Log in for availability
    Each - $189.98

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Syringe filter - ANOTOP 10
    Syringe filter - ANOTOP 10
    78172-066
    Non sterile, 0.02 µm membrane filter, Ø 10 mm, (1 piece)
    Log in for availability
    Each - $35.62

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software unlimitted
    Kalliope software unlimitted
    78172-078
    Measurement series, databases for solvents, materials and SOPs are all easily accessible;Results from separate experiments can be compared and analyzed, and presented in customizable reports and/or exported to an Excel file for further processing and analysis;In combination with KALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11 the software complies with the US FDA’s Regulation 21 CFR Part 11 concerning electronic records, audit trails and signatures; license for unlimited installations
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • C-vette for measuring particle size and transmittance
    C-vette for measuring particle size and transmittance
    78172-218
    Includes the capillary holder, 1 pack (25 pieces) of disposable capillary tubes and 1 hematocrit sealing wax
    Log in for availability
    Each - $1,181.97

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Quartz cuvette; Outer dimensions: 12.5×12.5×45 mm; light path 10 mm; Compatible with Univette (Cat No: 78172-090)
    Quartz cuvette; Outer dimensions: 12.5×12.5×45 mm; light path 10 mm; Compatible with Univette (Cat No: 78172-090)
    78172-070
    Log in for availability
    Each - $351.89

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Horiziontal and vertical polarization filter
    Horiziontal and vertical polarization filter
    78172-212
    Set of 1 horizontal and 1 vertical polarization filter; Can be applied to forward-, side- or backscatter angle
    Log in for availability
    Each - $441.48

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Vertical polarization filter PHARMA, 1 piece
    Vertical polarization filter PHARMA, 1 piece
    78172-214
    Can be applied to forward-, side- or backscatter angle
    Log in for availability
    Each - $551.59

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Fluorescence filter, 1 piece
    Fluorescence filter, 1 piece
    78172-208
    Can be applied to forward-, side- or backscatter angle
    Log in for availability
    Each - $441.48

Product Information

  • Catalog #
    Details
    Availability
    Price
  • OMEGA cuvette for ZETA and SIZE, box of 10 cuvettes, with 22 male Luer plugs
    OMEGA cuvette for ZETA and SIZE, box of 10 cuvettes, with 22 male Luer plugs
    78172-108
    Log in for availability
    Pack of 10 - $431.77

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Kalliope software professional
    Kalliope software professional
    78172-076
    One-page workflow displays all measurement details in one sight, including input parameters, results, and expert advice;Meets US FDA Regulation 21 CFR Part 11 standards for electronic records, audit trails, and signatures when used in conjunction with the KALLIOPE PROFESSIONAL SOFTWARE MODULE 21 CFR PART 11;Includes license for 3 activations (installations)
    Log in for availability
    Each

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Fluorescence filter PHARMA, 1 piece
    Fluorescence filter PHARMA, 1 piece
    78172-210
    Can be applied to forward-, side- or backscatter angle
    Log in for availability
    Each - $551.59

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Support plate (Pack of 3)
    Support plate (Pack of 3)
    78172-086
    For use with low-volume Quartz cuvette only (Cat. No. 78172-072)
    Log in for availability
    Pack of 3 - $38.86

Product Information

  • Catalog #
    Details
    Availability
    Price
  • UNIVETTE cell for DLS and ELS (Univette body)
    UNIVETTE cell for DLS and ELS (Univette body)
    78172-090
    Spacer for Univette; Low-volume accessory for Univette; Palladium electrodes; Quartz cuvette outer dimensions: 12.5×12.5×45 mm; light path 10 mm
    Log in for availability
    Each - $5,861.29

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Disposable cuvettes
    Disposable cuvettes
    78172-074
    Outer dimensions: 12×12×45 mm; light path 10 mm; pack of 100
    Log in for availability
    Pack of 100 - $57.21

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Starter kit Litesizer DLS 701/501 components
    Starter kit Litesizer DLS 701/501 components
    78172-062
    Log in for availability
    Each - $939.11

Product Information

  • Catalog #
    Details
    Availability
    Price
  • NIST molecular mass standard, 285 kDa
    NIST molecular mass standard, 285 kDa
    78172-080
    Log in for availability
    Each - $1,418.37

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Glass cuvette; Outer dimensions: 12.5×12.5×45 mm; light path 10 mm
    Glass cuvette; Outer dimensions: 12.5×12.5×45 mm; light path 10 mm
    78172-084
    Log in for availability
    Each - $145.72

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Low-volume quartz cuvette
    Low-volume quartz cuvette
    78172-072
    NOTE: The low-volume Quartz cuvette has a minimal volume of 15 µl, which can be reduced to 12 µl when used together with the support plate (Cat. No. 78172-086); Outer dimensions: 12.5×12.5×45 mm; light path 3 mm
    Log in for availability
    Each - $948.81

Product Information

  • Catalog #
    Details
    Availability
    Price
  • Cuvette rack for 10 mm cuvettes
    Cuvette rack for 10 mm cuvettes
    78172-064
    Log in for availability
    Each - $33.46

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.