Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Rapamycin

Rapamycin

Supplier:   G-Biosciences
Rapamycin
Supplier:  G-Biosciences
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Mode of action and Anti-microbial spectrum Rapamycin, a macrocyclic triene antibiotic, possesses immunosuppressant, anti-proliferative, and anti-cancer activity. Inhibition by rapamycin results in cell cycle arrest in G1. Rapamycin also has been shown to have a role in life span extension in mice.

  • Soluble in DMSO or Ethanol
Synonyms:  (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
  • CAS Number:53123-88-9
  • Formula:C₅₁H₇₉NO₁₃
  • Molecular Weight:914.17 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Vial
    100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $564.24
    $564.24
    Each (100mg)
       $564.24

    Supplier Number: RC-743
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Vial
    500 mg
    Log in for availability
    Log in for availability
    Each (500mg)
    Each (500mg) - $2,252.13
    $2,252.13
    Each (500mg)
       $2,252.13

    Supplier Number: RC-744
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.