Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Yeast Extract

Yeast Extract

Supplier:   G-Biosciences
Yeast Extract
Supplier:  G-Biosciences
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

An autolysate of yeast cells used in preparing microbiological culture media.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 71003-508
  • 71003-506
  • CAS No.:
  • 8013-01-2
  • 8013-01-2

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 71003-508
    Yeast extract
    Yeast extract
    500 g
    Log in for availability
    Log in for availability
    Each
    Each - $375.68
    $375.68
    Each
       $375.68

    Supplier Number: RC-117
    Specifications:
    More
    Specifications
    Cat. No.
    71003-508
    CAS No.
    8013-01-2
  • 71003-506
    Yeast extract
    Yeast extract
    100 g
    Log in for availability
    Log in for availability
    Each
    Each - $84.97
    $84.97
    Each
       $84.97

    Supplier Number: RC-116
    Specifications:
    More
    Specifications
    Cat. No.
    71003-506
    CAS No.
    8013-01-2

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.