VWR® Acidic Detergent
Acidic detergent, descaler, removes mineral deposits, hardness salts and lime scale, Phosphoric Acid, 1 gal.
Catalog # 78123-064
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
A concentrated acidic detergent and descaling agent for removing hard water salt deposits from waterbaths, ultrasonic tanks, automatic labware washing machines, autoclaves, stills, labware, glass, plastics, and metals.
- Dissolves calcium, magnesium, sodium, and iron deposits/just about every salt left behind from hard water
- Powerful enough to remove crystallized urine deposits from animal cages
Acidic detergent non-foaming formulation allows its use in automatic washers and spraying equipment.
Specifications
- Description:Acidic detergent, descaler, removes mineral deposits, hardness salts and lime scale
- Material:Phosphoric Acid
- Volume:1 gal.