Orders LinkContactUsLinkComponent
Microbial Count Plate
New Product
Microbial Count Plate
New Product
Certificates
Microbial Count Plate
21 Options Available Below
Product Details & Documents
GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.
  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required
GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.
Product Family Options
Product Information
  • Format
    Availability
    Price
  • 78172-934
    Pivot E. coli/Coliform count plate
    Pivot E. coli/Coliform count plate
    Dark purple red colony, 36±1 °C, 24±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $218.70
    $218.70
    Pack of 25
       $218.70
    Supplier Number: MCPG00325
    Specifications:
    New Product
    New Product
  • 78172-954
    Shigella count plate
    Shigella count plate
    Red colony, , 36±1 °C, 24±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $243.00
    $243.00
    Pack of 25
       $243.00
    Supplier Number: MCPG02525
    Specifications:
    New Product
    New Product
  • 78172-814
    Bacillus Psychrophilus count plate
    Bacillus Psychrophilus count plate
    Black, green colony, 36±1 °C, 26±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $243.00
    $243.00
    Pack of 25
       $243.00
    Supplier Number: MCPG01925
    Specifications:
    New Product
    New Product
  • 78172-698
    Aerobic bacillus count plate
    Aerobic bacillus count plate
    Red colony, 21±1 °C, 48-60h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $243.00
    $243.00
    Pack of 25
       $243.00
    Supplier Number: MCPG02425
    Specifications:
    New Product
    New Product
  • 78173-170
    Enterobacteriaceae EB count plate
    Enterobacteriaceae EB count plate
    Purple, red colony, 36±1 °C, 24±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $121.50
    $121.50
    Pack of 25
       $121.50
    Supplier Number: MCPG01625
    Specifications:
    New Product
    New Product
  • 78172-624
    Listeria count plate
    Listeria count plate
    Green colony, 36±1 °C, 24±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $145.80
    $145.80
    Pack of 25
       $145.80
    Supplier Number: MCPG00525
    Specifications:
    New Product
    New Product
  • 78173-394
    Coliform count plate
    Coliform count plate
    Green colony, 28±1 °C, 48-2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $60.75
    $60.75
    Pack of 25
       $60.75
    Supplier Number: MCPG01025
    Specifications:
    New Product
    New Product
  • 78172-262
    Lactic Acid Bacteria count plate
    Lactic Acid Bacteria count plate
    Purple, red colony, 36±1 °C, 48±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $291.60
    $291.60
    Pack of 25
       $291.60
    Supplier Number: MCPG01425
    Specifications:
    New Product
    New Product
  • 78173-060
    Enterococcus faecalis count plate
    Enterococcus faecalis count plate
    Red colony, 55±1 °C, 36±1h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $243.00
    $243.00
    Pack of 25
       $243.00
    Supplier Number: MCPG01825
    Specifications:
    New Product
    New Product
  • 78172-414
    Salmonella count plate
    Salmonella count plate
    Red colony, 36±1 °C, 48±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $121.50
    $121.50
    Pack of 25
       $121.50
    Supplier Number: MCPG01525
    Specifications:
    New Product
    New Product
  • 78173-198
    Staph count plate
    Staph count plate
    Red colony, 36±1 °C, 48±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $85.05
    $85.05
    Pack of 25
       $85.05
    Supplier Number: MCPG00225
    Specifications:
    New Product
    New Product
  • 78173-226
    Yeast and molds platecount plate
    Yeast and molds platecount plate
    Blue green colony, 36±1 °C, 24±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $60.75
    $60.75
    Pack of 25
       $60.75
    Supplier Number: MCPG00825
    Specifications:
    New Product
    New Product
  • 78173-204
    Vibrio parahaemolyticus count plate
    Vibrio parahaemolyticus count plate
    Gray colony with halo, 36±1 °C, 18-24h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $243.00
    $243.00
    Pack of 25
       $243.00
    Supplier Number: MCPG02825
    Specifications:
    New Product
    New Product
  • 78172-714
    Escherichia coli O157 count plate
    Escherichia coli O157 count plate
    Blue, green colony, 36±1 °C, 36h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $291.60
    $291.60
    Pack of 25
       $291.60
    Supplier Number: MCPG02725
    Specifications:
    New Product
    New Product
  • 78172-722
    Geobacillus stearothermophilus count plate
    Geobacillus stearothermophilus count plate
    Red colony, 36±1 °C, 18-24h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $243.00
    $243.00
    Pack of 25
       $243.00
    Supplier Number: MCPG01725
    Specifications:
    New Product
    New Product
  • 78172-270
    Pivot Coliform count plate
    Pivot Coliform count plate
    Blue purple colony, 36±1 °C, 24±2h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $218.70
    $218.70
    Pack of 25
       $218.70
    Supplier Number: MCPG00425
    Specifications:
    New Product
    New Product
  • 78172-942
    Aerobic count plate
    Aerobic count plate
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $60.75
    $60.75
    Pack of 25
       $60.75
    Supplier Number: MCPG00125
    Specifications:
    New Product
    New Product
  • 78172-248
    Psychrophilic Bacteria count plate
    Psychrophilic Bacteria count plate
    Red colony, 36±1 °C, 8-18h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $243.00
    $243.00
    Pack of 25
       $243.00
    Supplier Number: MCPG02925
    Specifications:
    New Product
    New Product
  • 78173-412
    E. coli/Coliform count plate
    E. coli/Coliform count plate
    Red colony, 36±1 °C, 18-24h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $60.75
    $60.75
    Pack of 25
       $60.75
    Supplier Number: MCPG01125
    Specifications:
    New Product
    New Product
  • 78173-424
    Bacillus Cereus count plate
    Bacillus Cereus count plate
    Bule purple red colony, 36±1 °C, 18-24h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $121.50
    $121.50
    Pack of 25
       $121.50
    Supplier Number: MCPG01325
    Specifications:
    New Product
    New Product
  • 78173-460
    Listeria monocytogenes count plate
    Listeria monocytogenes count plate
    Red colony, 36 ±1 °C, 24-48h incubation
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $291.60
    $291.60
    Pack of 25
       $291.60
    Supplier Number: MCPG02625
    Specifications:
    New Product