Orders LinkContactUsLinkComponent
Floor Prep Solutions
New Product
Supplier:   Brady Worldwide
Floor Prep Solutions
Supplier:  Brady Worldwide
New Product
Certificates
Floor Prep Solutions
Floor Prep Solutions
2 Options Available Below
Product Details & Documents
The ready-to-use floor cleaner for tape application. This purpose-built, eco-friendly cleaner prepares floors for optimal adhesion and long-lasting, professional markings.
  • Purpose-formulated solution
  • Ensures optimal adhesion
  • Green and Eco-friendly formula
  • Versatile application
Designed specifically to remove common dirt, dust, and light residue to get surfaces ready for tape. Maximizes floor tape adhesion and durability for lasting performance. Designed to be green and Eco-friendly, this safe and non-hazardous formula effectively cleans a wide range of floor types. Convenient jug size is ideal for larger cleaning tasks and frequent use.
Product Family Options
Product Information
  • Packaging
    Availability
    Price
  • 78495-352
    Floor prep solution
    Floor prep solution
    1 gallon jug
    Log in for availability
    Log in for availability
    Each
    Each - $88.54
    $88.54
    Each
       $88.54
    Supplier Number: 178946
    Specifications:
    New Product
    New Product
  • 78495-364
    Floor prep solution
    Floor prep solution
    4×1 gallon jug
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $351.81
    $351.81
    Pack of 4
       $351.81
    Supplier Number: 178948
    Specifications:
    New Product
    New Product