Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
EEB Expansion

EEB Expansion

EEB expansion, 6
Catalog # 78122-994
New Product
Supplier #:  EXP-EEB001
EEB Expansion
Catalog # 78122-994
New Product
CAS Number:  
EEB Expansion
EEB Expansion

EEB expansion, 6

CAS Number:  
Price
$37.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Additional Extraction EB (EEB) for use with the Ultra-long DNA sequencing kit V14 (SQK-ULK114).

Delivery information: This product is stable during shipping for seven days at temperatures up to 25 °C. Upon receipt, please store the expansion under the recommended conditions specified on the expansion label. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.

Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.

Specifications

Specifications

  • Description:
    EEB expansion
  • No. of reactions:
    6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.