Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Boc-hydrazide ≥99%

Boc-hydrazide ≥99%

New Product
Boc-hydrazide ≥99%
New Product
Certificates
Boc-hydrazide ≥99%
Boc-hydrazide ≥99%

6 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Boc-hydrazide is a versatile chemical compound widely utilized in organic synthesis and pharmaceutical research. Its unique structure, featuring a tert-butyl group, enhances its stability and solubility, making it an ideal candidate for various applications. This compound is particularly valuable in the synthesis of hydrazones and other nitrogen-containing heterocycles, which are essential in the development of agrochemicals and pharmaceuticals. Researchers appreciate Boc-hydrazide for its ability to facilitate the formation of peptide bonds, thereby playing a crucial role in peptide synthesis and drug development.

Additionally, Boc-hydrazide serves as a protecting group in organic reactions, allowing for selective functionalization of other reactive sites. Its ease of removal under mild conditions makes it a preferred choice among chemists. With its broad applicability in medicinal chemistry, including the design of inhibitors and other bioactive compounds, Boc-hydrazide stands out as a key reagent in the toolkit of researchers and industry professionals alike.

Store at 0 to 8 °C.

Synonyms:  tert-Butyl hydrazinecarboxylate
  • CAS Number:870-46-2
  • Formula:C₅H₁₂N₂O₂
  • MDL Number:MFCD00007593
  • Molecular Weight:132.2 g/mol
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $25.28
    $25.28
    Each(1/Itm/Bx)
       $25.28

    Supplier Number: 02286-25G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $55.60
    $55.60
    Each(1/Itm/Bx)
       $55.60

    Supplier Number: 02286-100G
    Specifications:
    New Product
    New Product
  • Plastic bottle
    10 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $3,664.88
    $3,664.88
    Each(1/Itm/Bx)
       $3,664.88

    Supplier Number: 02286-10KG
    Specifications:
    New Product
    New Product
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $419.56
    $419.56
    Each(1/Itm/Bx)
       $419.56

    Supplier Number: 02286-1KG
    Specifications:
    New Product
    New Product
  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $121.32
    $121.32
    Each(1/Itm/Bx)
       $121.32

    Supplier Number: 02286-250G
    Specifications:
    New Product
    New Product
  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,915.84
    $1,915.84
    Each(1/Itm/Bx)
       $1,915.84

    Supplier Number: 02286-5KG
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.