Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-Sorbitol 91-100.5% FCC

D-Sorbitol 91-100.5% FCC

Plastic bottle, 250 g
Catalog # CH02003-250G
New Product
Supplier #:  02003-250G
D-Sorbitol 91-100.5% FCC
Catalog # CH02003-250G
New Product
CAS Number:  50-70-4
D-Sorbitol 91-100.5% FCC
D-Sorbitol 91-100.5% FCC

Plastic bottle, 250 g

CAS Number:  50-70-4
Price
$20.22
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

D-Sorbitol is a versatile sugar alcohol widely recognized for its applications in the food, pharmaceutical, and cosmetic industries. This compound, also known as D-glucitol, is valued for its sweetening properties and serves as a low-calorie sugar substitute, making it an excellent choice for sugar-free products. In the pharmaceutical sector, D-Sorbitol is utilized as a humectant and stabilizer in formulations, enhancing the texture and moisture retention of creams and gels. Its ability to act as a bulking agent also makes it a preferred ingredient in various dietary supplements and functional foods.

Moreover, D-Sorbitol's unique characteristics, such as its non-cariogenic nature, make it particularly appealing for oral care products, where it helps to reduce the risk of dental cavities. Its applications extend to the production of biodegradable plastics and as a cryoprotectant in cell preservation, showcasing its potential in both environmental and biotechnological fields. With its multifunctional properties, D-Sorbitol stands out as a valuable ingredient for researchers and industry professionals seeking innovative solutions.

Store at RT.

Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:C₆H₁₄O₆
  • MDL Number:MFCD00004708
  • Molecular Weight:182.2 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.