Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Ethanolamine ≥99.6%

Glass bottle, 250 ml
Catalog # CH01731-250ML
New Product
 :  01731-250ML
Ethanolamine ≥99.6%
Catalog # CH01731-250ML
New Product
CAS Number:  141-43-5
Ethanolamine ≥99.6%
Ethanolamine ≥99.6%

Glass bottle, 250 ml

CAS Number:  141-43-5
Price
$20.22
Log in for availability
Each(250ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ethanolamine, also known as 2-aminoethanol, is a versatile compound widely utilized across various industries due to its unique properties. This colorless, viscous liquid is an important building block in the synthesis of surfactants, emulsifiers, and corrosion inhibitors. Ethanolamine is particularly valued in the production of personal care products, where it serves as a pH balancer and a stabilizing agent in formulations. Its ability to form stable emulsions makes it essential in cosmetics and skincare products, enhancing texture and performance.

In the industrial sector, ethanolamine plays a crucial role in the manufacturing of herbicides, pharmaceuticals, and detergents. Its effectiveness as a gas treatment agent for removing acidic gases, such as carbon dioxide and hydrogen sulfide, highlights its importance in environmental applications. Researchers also leverage ethanolamine in various chemical syntheses and analytical procedures, making it a staple in laboratories. With its multifunctional capabilities and broad applicability, ethanolamine stands out as a key ingredient for professionals seeking reliable and effective solutions in their formulations and processes.

Store at RT.

Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:C₂H₇NO
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Storage Temperature:Room Temperature
  • UN:2491
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.