Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Potassium bromide ≥99% (by titrimetric analysis) ACS

Plastic bottle, 100 g
Catalog # CH01245-100G
New Product
 :  01245-100G
Potassium bromide ≥99% (by titrimetric analysis) ACS
Catalog # CH01245-100G
New Product
CAS Number:  7758-02-3
Potassium bromide ≥99% (by titrimetric analysis) ACS
Potassium bromide ≥99% (by titrimetric analysis) ACS

Plastic bottle, 100 g

CAS Number:  7758-02-3
Price
$22.27
Log in for availability
Each(100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Potassium bromide is a versatile inorganic compound widely recognized for its applications in various industries, particularly in photography, pharmaceuticals, and analytical chemistry. This white crystalline salt is known for its high solubility in water, making it an ideal choice for use in aqueous solutions. In the photographic industry, potassium bromide serves as a key component in developing agents, enhancing image quality and contrast. In pharmaceuticals, it has been utilized as an anticonvulsant and sedative, showcasing its importance in medical formulations. Additionally, potassium bromide is employed in laboratories for its role as a reagent in various chemical reactions, contributing to its relevance in research settings.

The unique properties of potassium bromide, such as its stability and non-toxic nature, make it a preferred choice over similar compounds in many applications. Its ability to dissolve easily in water allows for convenient use in both industrial and research environments. Whether you are involved in photography, pharmaceuticals, or chemical analysis, potassium bromide offers reliable performance and versatility, making it an essential compound for professionals seeking quality and efficiency in their work.

Store at RT.

  • CAS Number:7758-02-3
  • Formula:KBr
  • MDL Number:MFCD00011358
  • Molecular Weight:119.1 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.