Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ascorbic acid 99.0-100.5% (by titrimetric analysis) USP

Ascorbic acid 99.0-100.5% (by titrimetric analysis) USP

Ascorbic acid 99.0-100.5% (by titrimetric analysis) USP
Certificates
Ascorbic acid 99.0-100.5% (by titrimetric analysis) USP
Ascorbic acid 99.0-100.5% (by titrimetric analysis) USP

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ascorbic acid, commonly known as Vitamin C, is a vital nutrient with a wide range of applications in both the food and pharmaceutical industries. This water-soluble vitamin is renowned for its antioxidant properties, which help protect cells from oxidative stress and support overall health. Ascorbic acid is widely used as a dietary supplement to boost immune function, enhance collagen synthesis, and promote skin health. In the food industry, it serves as a natural preservative and color stabilizer, ensuring the freshness and quality of various products, from juices to processed foods.

In addition to its health benefits, ascorbic acid plays a crucial role in research and development. It is utilized in analytical chemistry for the determination of metal ions and as a reducing agent in various chemical reactions. Its unique properties make it an essential component in cosmetic formulations, where it helps improve skin texture and reduce signs of aging. With its multifaceted applications, ascorbic acid is an indispensable compound for researchers and industry professionals seeking effective solutions in health, nutrition, and product formulation.

Store at RT.

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.1 g/mol
  • Storage Temperature:Room Temperature
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $20.22
    $20.22
    Each(1/Itm/Bx)
       $20.22

    Supplier Number: 01218-100G
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $96.04
    $96.04
    Each(1/Itm/Bx)
       $96.04

    Supplier Number: 01218-1KG
  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $35.38
    $35.38
    Each(1/Itm/Bx)
       $35.38

    Supplier Number: 01218-250G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.