Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Urea 99.0-100.5% (by titrimetric analysis) ACS

Urea 99.0-100.5% (by titrimetric analysis) ACS

Plastic bottle, 1 kg
Catalog # CH00561-1KG
New Product
Supplier #:  00561-1KG
Urea 99.0-100.5% (by titrimetric analysis) ACS
Catalog # CH00561-1KG
New Product
CAS Number:  57-13-6
Urea 99.0-100.5% (by titrimetric analysis) ACS
Urea 99.0-100.5% (by titrimetric analysis) ACS

Plastic bottle, 1 kg

CAS Number:  57-13-6
Price
$31.14
Log in for availability
Each(1kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Urea is a versatile organic compound widely recognized for its significant role in various industries, particularly in agriculture and pharmaceuticals. As a nitrogen-rich fertilizer, urea enhances soil fertility and promotes plant growth, making it an essential component in modern farming practices. Its high solubility allows for easy application and rapid absorption by plants, ensuring efficient nutrient delivery. In the pharmaceutical sector, urea is utilized in the formulation of creams and ointments due to its moisturizing properties, effectively treating dry skin conditions. Additionally, it serves as a key ingredient in the production of plastics, resins, and adhesives, showcasing its multifaceted applications across different fields.

With its ability to improve crop yields and its effectiveness in personal care products, urea stands out as a crucial compound for researchers and industry professionals alike. Its unique properties, such as low toxicity and biodegradability, further enhance its appeal, making it a preferred choice in sustainable practices. Whether for agricultural enhancement or as a functional ingredient in cosmetic formulations, urea continues to demonstrate its value and versatility in various applications.

Store at RT.

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:CH₄N₂O
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.