Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acetone ≥99.5% (by GC)

Acetone ≥99.5% (by GC)

New Product
Acetone ≥99.5% (by GC)
New Product
Certificates
Acetone ≥99.5% (by GC)
Acetone ≥99.5% (by GC)

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Acetone, also known as propan-2-one, is a highly versatile solvent widely utilized in various industries due to its excellent solvent properties and rapid evaporation rate. This colorless, flammable liquid is commonly employed in the production of plastics, fibers, and pharmaceuticals, making it an essential component in manufacturing processes. Acetone is particularly valued in laboratories for its ability to dissolve a wide range of organic compounds, facilitating various chemical reactions and extractions. Its effectiveness as a cleaning agent also makes it a popular choice for removing residues and contaminants from surfaces and equipment.

In addition to its industrial applications, acetone is frequently used in nail polish removers and personal care products, showcasing its relevance in everyday consumer goods. Researchers appreciate acetone for its low toxicity and high efficiency, allowing for safe handling in laboratory settings. With its unique combination of properties, acetone stands out as a crucial solvent that enhances productivity and efficiency across multiple sectors, from chemical manufacturing to cosmetics.

Store at room temperature.

Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:C₃H₆O
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Storage Temperature:Room Temperature
  • UN:1090
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    Each(1l)
    Each(1l) - $33.60
    $33.60
    Each(1l)
       $33.60

    Supplier Number: 02889-1L
    Specifications:
    New Product
    New Product
  • Plastic bottle
    205 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,686.66
    $1,686.66
    Each(1/Itm/Bx)
       $1,686.66

    Supplier Number: 02889-205L
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.