Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium hydroxide ≥95% (by titrimetric analysis) ACS

Calcium hydroxide ≥95% (by titrimetric analysis) ACS

Plastic bottle, 3 kg
Catalog # CH01229-2.5KG
New Product
Supplier #:  01229-2.5KG
Calcium hydroxide ≥95% (by titrimetric analysis) ACS
Catalog # CH01229-2.5KG
New Product
CAS Number:  1305-62-0
Calcium hydroxide ≥95% (by titrimetric analysis) ACS
Calcium hydroxide ≥95% (by titrimetric analysis) ACS

Plastic bottle, 3 kg

CAS Number:  1305-62-0
Price
$112.39
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
Product Details & Documents

Calcium hydroxide, also known as slaked lime or hydrated lime, is a versatile compound with significant applications across various industries. This inorganic compound is widely used in construction for its role in the production of lime mortar and plaster, providing excellent binding properties and durability. In the agricultural sector, calcium hydroxide is employed to improve soil quality by neutralizing acidity, enhancing nutrient availability, and promoting healthier crop yields. Additionally, it serves as a crucial ingredient in water treatment processes, where it aids in the removal of impurities and the stabilization of pH levels.

Beyond these applications, calcium hydroxide is also utilized in the food industry as a food additive and in the production of calcium-based products, such as calcium carbonate. Its unique properties, including high solubility and reactivity, make it an essential compound for researchers and industry professionals seeking effective solutions in their respective fields. With its broad range of uses and benefits, calcium hydroxide stands out as a reliable choice for enhancing product performance and ensuring quality across multiple applications.

Store at room temperature.

Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specification Test Results

  • Assay CaCO₃:
    ≤3.0%
  • Assay by titration:
    ≥95.0%

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    3 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.