Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-Mannitol 97-102% (dried basis) USP

D-Mannitol 97-102% (dried basis) USP

Plastic bottle, 1 kg
Catalog # CH00696-1KG
New Product
Supplier #:  00696-1KG
D-Mannitol 97-102% (dried basis) USP
Catalog # CH00696-1KG
New Product
CAS Number:  69-65-8
D-Mannitol 97-102% (dried basis) USP
D-Mannitol 97-102% (dried basis) USP

Plastic bottle, 1 kg

CAS Number:  69-65-8
Price
$39.49
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

D-Mannitol is a versatile sugar alcohol with a wide range of applications in the pharmaceutical, food, and cosmetic industries. Known for its sweet taste and low caloric content, D-Mannitol is commonly used as a sugar substitute in various food products, making it an ideal choice for those seeking healthier alternatives. In pharmaceuticals, it serves as an excipient in tablet formulations and is utilized for its osmotic diuretic properties, which can help in the treatment of conditions such as increased intracranial pressure and glaucoma. Additionally, D-Mannitol is employed in the formulation of intravenous solutions, enhancing the stability and solubility of active ingredients.

This compound also exhibits unique features such as its ability to protect proteins and enzymes during freeze-drying processes, making it invaluable in biopharmaceutical applications. Its non-toxic nature and compatibility with a variety of formulations further enhance its appeal to researchers and industry professionals. With its multifunctional properties and practical applications, D-Mannitol stands out as a crucial ingredient in both health and food sectors, offering significant benefits over traditional sugar substitutes.

Store at room temperature.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:C₆H₁₄O₆
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.