Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ethidium bromide ≥99% (by HPLC)

Ethidium bromide ≥99% (by HPLC)

Plastic bottle, 1 g
Catalog # CH00666-1G
New Product
Supplier #:  00666-1G
Ethidium bromide ≥99% (by HPLC)
Catalog # CH00666-1G
New Product
CAS Number:  1239-45-8
Ethidium bromide ≥99% (by HPLC)
Ethidium bromide ≥99% (by HPLC)

Plastic bottle, 1 g

CAS Number:  1239-45-8
Price
$22.27
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ethidium bromide is a powerful intercalating agent widely utilized in molecular biology and biochemistry for its ability to bind to nucleic acids. This compound is particularly renowned for its application in gel electrophoresis, where it serves as a fluorescent stain for DNA and RNA visualization. When exposed to ultraviolet light, ethidium bromide emits a bright orange fluorescence, making it an essential tool for researchers analyzing genetic material. Its effectiveness in detecting nucleic acids has made it a staple in laboratories for applications such as cloning, sequencing, and PCR analysis.

In addition to its role in nucleic acid visualization, ethidium bromide is also employed in various research settings to study cell viability and apoptosis. Its unique properties allow researchers to assess the integrity of cellular membranes, providing insights into cellular health and function. While handling ethidium bromide requires caution due to its mutagenic properties, its benefits in molecular research are unparalleled, making it a vital compound for professionals in the life sciences.

Store at room temperature.

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.32 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.