Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Uridine ≥99% (by HPLC)

Uridine ≥99% (by HPLC)

Plastic bottle, 100 g
Catalog # CH00339-100G
New Product
Supplier #:  00339-100G
Uridine ≥99% (by HPLC)
Catalog # CH00339-100G
New Product
CAS Number:  58-96-8
Uridine ≥99% (by HPLC)
Uridine ≥99% (by HPLC)

Plastic bottle, 100 g

CAS Number:  58-96-8
Price
$75.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Uridine is a naturally occurring nucleoside that plays a crucial role in various biological processes, particularly in RNA synthesis and metabolism. It is recognized for its potential applications in the fields of neuroscience and cellular biology, where it is utilized to support cognitive function and promote neuronal health. Researchers have explored its use in enhancing memory and learning capabilities, making it a valuable compound in neuropharmacology. Additionally, Uridine has been studied for its potential benefits in treating conditions such as depression and cognitive decline, showcasing its relevance in therapeutic applications.

In the pharmaceutical industry, Uridine is also significant for its role in the synthesis of nucleotides, which are essential for DNA and RNA production. This makes it an important compound in biochemistry and molecular biology research. Its unique properties, such as being a precursor to important biomolecules, highlight its versatility and importance in various scientific studies. With its growing recognition in both research and therapeutic contexts, Uridine stands out as a compound of interest for professionals seeking to explore its multifaceted applications.

Store at room temperature.

Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.2 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specification Test Results

  • Purity by HPLC :
    ≥99%
  • Purity by UV:
    98 - 102%

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.