Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-Biotin 97.5-102.0% USP

D-Biotin 97.5-102.0% USP

New Product
D-Biotin 97.5-102.0% USP
New Product
Certificates
D-Biotin 97.5-102.0% USP
D-Biotin 97.5-102.0% USP

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

D-Biotin, also known as Vitamin H or Coenzyme R, is a vital water-soluble B-vitamin that plays a crucial role in various metabolic processes. This compound is essential for the synthesis of fatty acids, amino acids, and glucose, making it indispensable in the fields of nutrition and biochemistry. D-Biotin is widely utilized in dietary supplements and functional foods due to its ability to support healthy skin, hair, and nails, as well as its importance in energy metabolism. Researchers and industry professionals appreciate its role in promoting cellular health and its potential therapeutic applications in conditions such as biotin deficiency and metabolic disorders.

In addition to its nutritional benefits, D-Biotin is increasingly being explored for its applications in cosmetic formulations, where it is valued for its skin-conditioning properties. Its unique structure allows for effective incorporation into various formulations, enhancing product efficacy. With its proven benefits and versatile applications, D-Biotin stands out as a key ingredient for those looking to improve health and wellness through scientifically-backed solutions.

Store at 0 to 8 °C.

Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1g)
    Each(1g) - $18.53
    $18.53
    Each(1g)
       $18.53

    Supplier Number: 00033-1G
    Specifications:
    New Product
    New Product
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $126.38
    $126.38
    Each(1/Itm/Bx)
       $126.38

    Supplier Number: 00033-25G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • Plastic bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $40.44
    $40.44
    Each(1/Itm/Bx)
       $40.44

    Supplier Number: 00033-5G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.