Orders LinkContactUsLinkComponent
Closeable Wipe Holder
Closeable Wipe Holder
Closeable wipe holder, Stainless steel, 10.13×8.0×9.98"
Catalog # 78469-870
New Product
Supplier:   REYTEK EQUIPMENT
Supplier #:  REY1909-500
Closeable Wipe Holder
Catalog # 78469-870
Supplier:  REYTEK EQUIPMENT
New Product
Closeable Wipe Holder
Closeable Wipe Holder
Closeable wipe holder, Stainless steel, 10.13×8.0×9.98"
Price
$793.20
Log in for availability
Each
Product Details & Documents
Protect and organize wipes in cleanrooms with this closeable 316 stainless steel holder. The all-welded, hygienic design is easy to clean and built to endure harsh conditions.
  • Superior 316 stainless steel provides enhanced corrosion resistance
  • Smooth surfaces for effortless cleaning
  • Secure closeable design protects wipes from contamination
Ideal for cleanrooms, labs, and production areas, ensuring wipes are always accessible and contamination-free.
Documents
Specifications
  • Description:
    Closeable wipe holder
  • Material:
    Stainless steel
  • W×D×H:
    10.13×8.0×9.98"