Dextran, 200,000
Dextran is a polysaccharide composed mainly of glucose linked by α-(1-6) bonds. This product has an average molecular weight of 180000 to 210000 and is used for leukocyte separation, as a thickening agent, and as a viscosity stabilizer.
: For research use only (RUO).
- :9004-54-0
- :(C₆H₁₀O₅)n
- :Average 180.000 - 210.000
- :Ambient
- Size:100 g