Document Holder
Document holder, Stainless steel, 9.2W×2.9D×12.5"H
Catalog # 78469-860
New Product
Supplier: REYTEK EQUIPMENT
Supplier #: REY1540
Jump to:
Product Details & Documents
Keep cleanrooms organized with this rugged stainless steel document holder. Made from 304 brushed stainless steel with all-welded construction, it’s durable, hygienic, and easy to clean. The sleek design ensures compliance with strict standards while maintaining a polished look.
- Durable and hygienic material is resistant to corrosion and simple to sanitize Space-saving design; compact dimensions fit seamlessly into any cleanroom setup Brushed finish adds a modern, high-quality professional appearance
Ideal for gowning rooms, production areas, and controlled environments, this document holder keeps SOPs, logs, and critical paperwork secure and accessible.
Documents
Cutsheet 78469-860 REY1540.pdf
Technical Drawing
Specifications
- Description:Document holder
- Dimensions:9.2W×2.9D×12.5"H
- Material:Stainless steel