Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Adenosine 98-102% (dried basis) USP

Plastic bottle, 25 g
Catalog # CH00007-25G
New Product
 :  00007-25G
Adenosine 98-102% (dried basis) USP
Catalog # CH00007-25G
New Product
CAS Number:  58-61-7
Adenosine 98-102% (dried basis) USP
Adenosine 98-102% (dried basis) USP

Plastic bottle, 25 g

CAS Number:  58-61-7
Price
$25.28
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Adenosine is a naturally occurring nucleoside that plays a crucial role in various biological processes, making it a valuable compound in both research and industry. Known for its involvement in energy transfer and signal transduction, adenosine is a key player in cellular metabolism and is essential for the synthesis of ATP (adenosine triphosphate). Its unique properties allow it to act as a signaling molecule in the cardiovascular system, influencing heart rate and blood flow, which has led to its exploration in therapeutic applications for conditions such as arrhythmias and heart failure.

In addition to its physiological significance, adenosine is widely used in the pharmaceutical industry for the development of drugs targeting neurological disorders, as it has neuroprotective effects and can modulate neurotransmitter release. Researchers also utilize adenosine in various biochemical assays and studies related to cellular signaling pathways. Its versatility and essential role in numerous biological functions make adenosine a compound of great interest for professionals in biochemistry, pharmacology, and related fields.

Shipping temperature: Store at 0 to 8 °C

Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.