Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
AquaBlot™ 10× High Efficiency Transfer Buffer

AquaBlot™ 10× High Efficiency Transfer Buffer

1 L
Catalog # 77973-793
New Product
Supplier #:  015-26213
AquaBlot™ 10× High Efficiency Transfer Buffer
Catalog # 77973-793
New Product
CAS Number:  
AquaBlot™ 10× High Efficiency Transfer Buffer
AquaBlot™ 10× High Efficiency Transfer Buffer

1 L

CAS Number:  
Price
$191.66
Log in for availability
Each(1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

AquaBlot™ 10× High efficiency transfer buffer is a high-performance liquid buffer used for protein transfer in blotting applications. This is a 10× transfer buffer superior in transfer efficiency to the Towbin buffer (Tris-glycine-methanol).Dilute 10 times with deionized water before use. No methanol needs to be added. Methanol is necessary for wetting a PVDF membrane.

  • Higher transfer efficiency than conventional methods
  • Simply change the transfer buffer
  • No methanol addition required

Caution: For research use only (RUO).

Specifications

Specifications

  • Size:
    1 L
  • Cat. No.:
    77973-793
  • Storage Temperature:
    RT

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.