ScreenFect™ A
Product Details & Documents
ScreenFect™ A is a low-toxicity transfection reagent made of novel cationic lipids, effective for DNA and siRNA delivery across various eukaryotic cell types - including common cell lines, stem cells, blood cells, microglia, and primary cultures. It can be directly added to media containing antibiotics and serum without needing medium change post-transfection, and contains no harmful substances.
- High transfection efficiency
- Non-hazardous
- Includes dilution buffer
- Easy to use
- Suitable for various mammalian cell lines
Caution: For research use only (RUO).
Specifications
Specifications
- Size:1 ml
- Cat. No.:77974-908
- Storage Temperature:Refrigerated (at 2...10 °C storage)