Orders LinkContactUsLinkComponent
β-Cyclodextrine ≥98% cell culture reagent
New Product
β-Cyclodextrine ≥98% cell culture reagent
New Product
β-Cyclodextrine ≥98% cell culture reagent
β-Cyclodextrin(β-CD) has been used in the synthesis of mono-6-tosyl-CD. It has also been used as a constituent in the serum-free medium to assess the involvement of cholesterol-dependent mechanisms in the uptake of nanoparticles by rat basophilic leukemia cells.
 :  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  •  :7585-39-9
  •  :C₄₂H₇₀O₃₅
  •  :MFCD00078139
  •  :1134.98 g/mol
  •  :Ambient
  •  :231-493-2
Product Family Options
  • Pack type
    Size
  • Plastic bottle
    100 g
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $140.00
    $140.00
    Each(1/Itm/Bx)
       $140.00
     : C118530-100G
    New Product
    New Product
  • Plastic bottle
    25 g
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $59.90
    $59.90
    Each(1/Itm/Bx)
       $59.90
     : C118530-25G
    New Product
    New Product
  • Plastic bottle
    5 g
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $17.90
    $17.90
    Each(1/Itm/Bx)
       $17.90
     : C118530-5G