β-Cyclodextrine ≥96%
Bottle, 250 g
Catalog # ALC104385-250G
New Product
Supplier: Aladdin Scientific
Supplier #: C104385-250G
β-Cyclodextrine ≥96%
Catalog # ALC104385-250G
Supplier: Aladdin Scientific
New Product
CAS Number: 7585-39-9
Jump to:
Product Details & Documents
Useful for forming water-soluble complexes for fluorescent labeling of proteins.
Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
- CAS Number:7585-39-9
- Formula:C₄₂H₇₀O₃₅
- MDL Number:MFCD00078139
- Molecular Weight:1134.98 g/mol
- Storage Temperature:Ambient
- EINECS:231-493-2
Specifications
Specifications
- Pack type:Bottle
- Size:250 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



