Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium propionate ≥99%

Calcium propionate ≥99%

Bottle, 100 g
  ALC492256-100G
 
 
New Product
Supplier #:  C492256-100G
Calcium propionate ≥99%
  ALC492256-100G
New Product
 :  4075-81-4
Calcium propionate ≥99%

Bottle, 100 g

 :  4075-81-4
 
$13.90
   
Each(1/Itm/Bx)
 
   
 
 :  Calcium dipropionate, Propionic acid hemicalcium salt, Propionic acid calcium salt
  •  :4075-81-4
  •  :C₆H₁₀CaO₄
  •  :MFCD00036137
  •  :186.22 g/mol
  •  :Ambient
  •  :223-795-8
 

 

  • Pack type:
    Bottle
  • Size:
    100 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.