Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-(+)-Biotin ≥98%, Moligand™

D-(+)-Biotin ≥98%, Moligand™

Supplier:   Aladdin Scientific
D-(+)-Biotin ≥98%, Moligand™
Supplier:  Aladdin Scientific
Certificates
D-(+)-Biotin ≥98%, Moligand™

6 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Essential vitamin that is important for amino acid and energy metabolism. Application: Biotin has been used for testing the functionality of the Avidin-displaying phages.The product has also been used for labeling the swine anti-rabbit IgG antibody and peroxidase. It has been used for culturing of oligodendrocytes. It has been used as a vitamin supplement for the growth of Bacillus species. It has been used for blocking endogenous biotin during immunohistology procedures.

Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $308.00
    $308.00
    Each(1/Itm/Bx)
       $308.00

    Supplier Number: B105433-100G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    10 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $41.90
    $41.90
    Each(1/Itm/Bx)
       $41.90

    Supplier Number: B105433-10G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $16.90
    $16.90
    Each(1/Itm/Bx)
       $16.90

    Supplier Number: B105433-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $101.00
    $101.00
    Each(1/Itm/Bx)
       $101.00

    Supplier Number: B105433-25G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    Each(500g)
    Each(500g) - $774.00
    $774.00
    Each(500g)
       $774.00

    Supplier Number: B105433-500G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $34.90
    $34.90
    Each(1/Itm/Bx)
       $34.90

    Supplier Number: B105433-5G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.