Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
γ-Cyclodextrine ≥98%

γ-Cyclodextrine ≥98%

Irritant
New Product
Supplier:   Aladdin Scientific
γ-Cyclodextrine ≥98%
Supplier:  Aladdin Scientific
New Product
Certificates
γ-Cyclodextrine ≥98%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:241-482-4
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $217.00
    $217.00
    Each(1/Itm/Bx)
       $217.00

    Supplier Number: C103968-500G
    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $9.90
    $9.90
    Each(1/Itm/Bx)
       $9.90

    Supplier Number: C103968-5G
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.