Chlorophenol red indicator
New Product
Supplier: Aladdin Scientific
Jump to:
About this item
Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Number:4430-20-0
- Formula:C₁₉H₁₂Cl₂O₅S
- MDL Number:MFCD00005877
- Molecular Weight:423.27 g/mol
- Storage Temperature:Ambient
- EINECS:224-619-2
3 Options Available Below
Product Details & Documents
Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Number:4430-20-0
- Formula:C₁₉H₁₂Cl₂O₅S
- MDL Number:MFCD00005877
- Molecular Weight:423.27 g/mol
- Storage Temperature:Ambient
- EINECS:224-619-2
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: C104980-1GSpecifications:New ProductNew Product
- Supplier Number: C104980-25GSpecifications:New ProductNew Product
- Supplier Number: C104980-5GSpecifications:New ProductNew Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



