Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Behenic acid ≥85%, Technical Grade

Behenic acid ≥85%, Technical Grade

Bottle, 100 g
Catalog # ALB105674-100G
Supplier:   Aladdin Scientific
Supplier #:  B105674-100G
Behenic acid ≥85%, Technical Grade
Catalog # ALB105674-100G
Supplier:  Aladdin Scientific
CAS Number:  112-85-6
Behenic acid ≥85%, Technical Grade

Bottle, 100 g

CAS Number:  112-85-6
Price
$22.90
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.58 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-010-8
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.