Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Bismuth(III) carbonate basic ≥90%, Analytical reagent (AR)

Plastic bottle, 100 g
Catalog # ALB112505-100G
Supplier:   Aladdin Scientific
 :  B112505-100G
Bismuth(III) carbonate basic ≥90%, Analytical reagent (AR)
Catalog # ALB112505-100G
Supplier:  Aladdin Scientific
CAS Number:  5892-10-4
Bismuth(III) carbonate basic ≥90%, Analytical reagent (AR)

Plastic bottle, 100 g

CAS Number:  5892-10-4
Price
$22.90
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS Number:5892-10-4
  • Formula:CBi₂O₅
  • MDL Number:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Storage Temperature:Ambient
  • EINECS:227-567-9
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.