Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Copper(II) oxide ≥99.5%, powder

Glass bottle, 100 g
Catalog # ALC490141-100G
Environment
New Product
Supplier:   Aladdin Scientific
 :  C490141-100G
Copper(II) oxide ≥99.5%, powder
Catalog # ALC490141-100G
Supplier:  Aladdin Scientific
New Product
CAS Number:  1317-38-0
Copper(II) oxide ≥99.5%, powder

Glass bottle, 100 g

CAS Number:  1317-38-0
Price
$58.90
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Slightly hygroscopic, soluble in dilute acid, alkali cyanide or ammonium carbonate solution, slowly soluble in ammonia, almost insoluble in water and alcohol. Copper oxide can be made into powder, filiform and other different shapes as needed.

Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • Molecular Weight:79.55 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-269-1
  • UN:3077
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.