Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Copper ≥99.999%, powder, PrimorTrace™

Copper ≥99.999%, powder, PrimorTrace™

Environment
New Product
Supplier:   Aladdin Scientific
Copper ≥99.999%, powder, PrimorTrace™
Supplier:  Aladdin Scientific
New Product
Certificates
Copper ≥99.999%, powder, PrimorTrace™

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • Molecular Weight:63.55 g/mol
  • Storage Temperature:Ambient
  • UN:3089
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    10 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $574.00
    $574.00
    Each(1/Itm/Bx)
       $574.00

    Supplier Number: C434853-10G
    Specifications:
    New Product
    New Product
  • Plastic bottle
    50 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,510.00
    $1,510.00
    Each(1/Itm/Bx)
       $1,510.00

    Supplier Number: C434853-50G
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.