Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Cedrol ≥98%

Environment
New Product
Supplier:   Aladdin Scientific
Cedrol ≥98%
Supplier:  Aladdin Scientific
New Product
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

(+)-Cedrol is a crystalline hydrated product of α-cedrene, which is a sesquiterpene found in cedar-wood oil. It can be used as a fragrance ingredient in cosmetics, shampoos, and soaps as well as in non-cosmetic products such as cleaners and detergents. (±) Cedrol can be synthesized by intramolecular Diels-Alder reaction of alkyl cyclopentadiene. Product Application(+)-Cedrol can be used as a starting material for the preparation of cedryl acetate by acetylation using acetic anhydride in the presence of an acid catalyst. It can also be incorporated as a precursor for the total synthesis of rare illicium sesquiterpene (+)-pseudoanisatin via selective C-H bond functionalization.

Synonyms:  (+)-Cedrol, (3R,3aS,6R,7R,8aS)-3,6,8,8-Tetramethyloctahydro-1H-3a,7-methanoazulen-6-ol
  • CAS Number:77-53-2
  • Formula:C₁₅H₂₆O
  • MDL Number:MFCD00062952
  • Molecular Weight:222.37 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $18.90
    $18.90
    Each(1/Itm/Bx)
       $18.90

    Supplier Number: C154086-25G
    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $9.90
    $9.90
    Each(1/Itm/Bx)
       $9.90

    Supplier Number: C154086-5G
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.